missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide, 99%

Catalog No. AAL0450322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0450322 100 g
AAL0450336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0450322 Supplier Thermo Scientific Chemicals Supplier No. L0450322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 617-84-5 | C5H11NO | 101.15 g/mol

N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

Solubility
Miscible with acetone, benzene, alcohol and ether.  

Notes
Incompatible with strong oxidizing agents. Avoid heat, flames and spark.                      

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

Density 0.91
Boiling Point 178°C
Flash Point 65°C (149°F)
Odor Amine-like
Refractive Index 1.434
Quantity 100 g
Beilstein 1209392
Solubility Information Miscible with acetone,benzene,alcohol and ether.
Formula Weight 101.15
Percent Purity 99%
Chemical Name or Material N,N-Diethylformamide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 210-533-2
RTECSNumber LQ1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only