missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylethylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 146°C |
| Quantity | 500 mL |
| UN Number | 2685 |
| Formula Weight | 116.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | N,N-Diethylethylenediamine |
Safety and Handling
| EINECSNumber | (2)-0158 |
| RTECSNumber | KV3500000 |
| TSCA | Yes |