missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethylenediamine 98.0+%, TCI America™

Catalog No. D050525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D050525ML 25 mL
D0505500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D050525ML Supplier TCI America Supplier No. D050525ML

Chemical Identifiers

CAS 100-36-7
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008176
InChI Key UDGSVBYJWHOHNN-UHFFFAOYSA-N
Synonym n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine
PubChem CID 60993
IUPAC Name N',N'-diethylethane-1,2-diamine
SMILES CCN(CC)CCN

Specifications

Color Yellow
Boiling Point 146°C
Quantity 25 mL
UN Number 2685
Formula Weight 116.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylethylenediamine
EINECSNumber (2)-0158
RTECSNumber KV3500000
TSCA Yes