missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethylenediamine, 98+%

Catalog No. p-109171
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150701000 100 mL Glass bottle
AC150705000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC150701000 Supplier Thermo Scientific Chemicals Supplier No. 150701000

CAS: 100-36-7 | C6H16N2 | 116.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-36-7
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00008176
InChI Key UDGSVBYJWHOHNN-UHFFFAOYSA-N
Synonym n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine
PubChem CID 60993
IUPAC Name N',N'-diethylethane-1,2-diamine
SMILES CCN(CC)CCN

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 145°C to 147°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4350 to 1.437
Linear Formula (C2H5)2NCH2CH2NH2
Quantity 100 mL
Beilstein 04, 251
Specific Gravity 0.82
Solubility Information Solubility in water: miscible. Other solubilities: soluble in ether,ethanol,toluene,ccl4
Formula Weight 116.21
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material N, N-Diethylethylenediamine, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
EINECSNumber 202-844-7
RTECSNumber KV3500000
TSCA TSCA

RUO – Research Use Only