missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyldodecanamide 98.0+%, TCI America™

Catalog No. p-7126717
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D140725ML 25 mL
D1407500ML 500 mL
2 options

Catalog No. D140725ML

Supplier: TCI America D140725ML

Only null left

Chemical Identifiers

CAS 3352-87-2
Molecular Formula C16H33NO
Molecular Weight (g/mol) 255.446
MDL Number MFCD00008962
InChI Key CWNSVVHTTQBGQB-UHFFFAOYSA-N
Synonym dodecanamide, n,n-diethyl, diethyllauramide, n,n-diethyllauramide, n,n-diethyllaurylamide, nopcogen 14-l, unii-0aj0n90byy, 0aj0n90byy, dodecanamide,n-diethyl, n,n-diethyl lauramide, n-lauroyldiethylamine
PubChem CID 18783
IUPAC Name N,N-diethyldodecanamide
SMILES CCCCCCCCCCCC(=O)N(CC)CC

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
Formula Weight 255.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyldodecanamide
EINECSNumber (2)-0806
RTECSNumber RB4200000
TSCA Yes