missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyldodecanamide 98.0+%, TCI America™

Catalog No. D140725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D140725ML 25 mL
D1407500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D140725ML Supplier TCI America Supplier No. D140725ML

Chemical Identifiers

CAS 3352-87-2
Molecular Formula C16H33NO
Molecular Weight (g/mol) 255.446
MDL Number MFCD00008962
InChI Key CWNSVVHTTQBGQB-UHFFFAOYSA-N
Synonym dodecanamide, n,n-diethyl, diethyllauramide, n,n-diethyllauramide, n,n-diethyllaurylamide, nopcogen 14-l, unii-0aj0n90byy, 0aj0n90byy, dodecanamide,n-diethyl, n,n-diethyl lauramide, n-lauroyldiethylamine
PubChem CID 18783
IUPAC Name N,N-diethyldodecanamide
SMILES CCCCCCCCCCCC(=O)N(CC)CC

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
Formula Weight 255.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyldodecanamide
EINECSNumber (2)-0806
RTECSNumber RB4200000
TSCA Yes