missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylcyclohexylamine 98.0+%, TCI America™

Catalog No. D0484500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0484500ML 500 mL
D048425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0484500ML Supplier TCI America Supplier No. D0484500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 91-65-6
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.29
MDL Number MFCD00003847
InChI Key CIXSDMKDSYXUMJ-UHFFFAOYSA-N
PubChem CID 61571
IUPAC Name N,N-diethylcyclohexanamine
SMILES CCN(CC)C1CCCCC1

Specifications

Color Yellow
Boiling Point 193°C
Quantity 500 mL
UN Number 2734
Formula Weight 155.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylcyclohexylamine
EINECSNumber (3)-2274
RTECSNumber GU5830125
TSCA Yes