missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylchloroacetamide 98.0+%, TCI America™
$70.84 - $639.37
Chemical Identifiers
| CAS | 2315-36-8 |
|---|---|
| Molecular Formula | C6H12ClNO |
| Molecular Weight (g/mol) | 149.618 |
| MDL Number | MFCD00000928 |
| InChI Key | CQQUWTMMFMJEFE-UHFFFAOYSA-N |
| Synonym | n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide |
| PubChem CID | 16844 |
| IUPAC Name | 2-chloro-N,N-diethylacetamide |
| SMILES | CCN(CC)C(=O)CCl |
Chemical Identifiers
| 2315-36-8 | |
| 149.618 | |
| CQQUWTMMFMJEFE-UHFFFAOYSA-N | |
| 16844 | |
| CCN(CC)C(=O)CCl |
| C6H12ClNO | |
| MFCD00000928 | |
| n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide | |
| 2-chloro-N,N-diethylacetamide |
Specifications
| 2315-36-8 | |
| 150°C | |
| MFCD00000928 | |
| 2810 | |
| CQQUWTMMFMJEFE-UHFFFAOYSA-N | |
| 2-chloro-N,N-diethylacetamide | |
| 16844 | |
| ≥98.0% (GC) | |
| N,N-Diethylchloroacetamide |
| Yellow | |
| C6H12ClNO | |
| 25 g | |
| n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide | |
| CCN(CC)C(=O)CCl | |
| 149.618 | |
| 149.62 | |
| Liquid |
Safety and Handling
RTECSNumber : AB5425000
TSCA : Yes