missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylaniline 99.0+%, TCI America™

Catalog No. p-7126235
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D047525ML 25 mL
D0475500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D047525ML Supplier TCI America Supplier No. D047525ML
Only null left

Chemical Identifiers

CAS 91-66-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00009042,MFCD31699978
InChI Key GGSUCNLOZRCGPQ-UHFFFAOYSA-N
Synonym diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine
PubChem CID 7061
IUPAC Name N,N-diethylaniline
SMILES CCN(CC)C1=CC=CC=C1

Specifications

Melting Point -38°C
Color Yellow
Boiling Point 216°C
Quantity 25 mL
UN Number 2432
Formula Weight 149.24
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylaniline
EINECSNumber (3)-0114
RTECSNumber BX3400000
TSCA Yes