missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylacrylamide (stabilized with MEHQ) 98.0+%, TCI America™
$78.29 - $335.52
Chemical Identifiers
| CAS | 2675-94-7 |
|---|---|
| Molecular Formula | C7H13NO |
| Molecular Weight (g/mol) | 127.187 |
| MDL Number | MFCD00078408 |
| InChI Key | OVHHHVAVHBHXAK-UHFFFAOYSA-N |
| PubChem CID | 17583 |
| IUPAC Name | N,N-diethylprop-2-enamide |
| SMILES | CCN(CC)C(=O)C=C |
Chemical Identifiers
| 2675-94-7 | |
| 127.187 | |
| OVHHHVAVHBHXAK-UHFFFAOYSA-N | |
| N,N-diethylprop-2-enamide |
| C7H13NO | |
| MFCD00078408 | |
| 17583 | |
| CCN(CC)C(=O)C=C |
Specifications
| 2675-94-7 | |
| 95°C | |
| MFCD00078408 | |
| OVHHHVAVHBHXAK-UHFFFAOYSA-N | |
| N,N-diethylprop-2-enamide | |
| 17583 | |
| ≥98.0% (GC,T) | |
| N,N-Diethylacrylamide (stabilized with MEHQ) |
| Yellow | |
| C7H13NO | |
| 25 g | |
| CCN(CC)C(=O)C=C | |
| 127.187 | |
| 127.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1017
RTECSNumber : AS3471000
TSCA : No