missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylacrylamide (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. D3737500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3737500G 500 g
D373725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3737500G Supplier TCI America Supplier No. D3737500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2675-94-7
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00078408
InChI Key OVHHHVAVHBHXAK-UHFFFAOYSA-N
PubChem CID 17583
IUPAC Name N,N-diethylprop-2-enamide
SMILES CCN(CC)C(=O)C=C

Specifications

Color Yellow
Boiling Point 95°C
Quantity 500 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylacrylamide (stabilized with MEHQ)
EINECSNumber (2)-1017
RTECSNumber AS3471000
TSCA No