missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylacetoacetamide 98.0+%, TCI America™

Catalog No. D137125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D137125G 25 g
D1371500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D137125G Supplier TCI America Supplier No. D137125G

Chemical Identifiers

CAS 2235-46-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00026728
InChI Key NTMXFHGYWJIAAE-UHFFFAOYSA-N
Synonym n,n-diethylacetoacetamide, n,n-diethyl-3-oxobutyramide, diethylacetoacetamide, butanamide, n,n-diethyl-3-oxo, acetoacetamide, n,n-diethyl, 1-diethylcarbamoyl-2-propanone, n,n-diethylacetylacetamide, acetoacetdiethylamide, diethylamid kyseliny acetoctove, diethylamid kyseliny acetoctove czech
PubChem CID 16699
IUPAC Name N,N-diethyl-3-oxobutanamide
SMILES CCN(CC)C(=O)CC(=O)C

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylacetoacetamide
EINECSNumber (2)-3338
RTECSNumber AK4025000
TSCA Yes