missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylacetoacetamide, 97%

Catalog No. p-7027344
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2401918 50 g
AAB2401930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2401918 Supplier Thermo Scientific Chemicals Supplier No. B2401918
Only null left

CAS: 2235-46-3 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2235-46-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00026728
InChI Key NTMXFHGYWJIAAE-UHFFFAOYSA-N
Synonym n,n-diethylacetoacetamide, n,n-diethyl-3-oxobutyramide, diethylacetoacetamide, butanamide, n,n-diethyl-3-oxo, acetoacetamide, n,n-diethyl, 1-diethylcarbamoyl-2-propanone, n,n-diethylacetylacetamide, acetoacetdiethylamide, diethylamid kyseliny acetoctove, diethylamid kyseliny acetoctove czech
PubChem CID 16699
IUPAC Name N,N-diethyl-3-oxobutanamide
SMILES CCN(CC)C(=O)CC(=O)C

Specifications

Melting Point <-73°C
Density 0.99
Boiling Point 213°C to 218°C
Flash Point 94°C (201°F)
Refractive Index 1.472
Quantity 50 g
Beilstein 636486
Formula Weight 157.2
Percent Purity 97%
Chemical Name or Material N,N-Diethylacetoacetamide
EINECSNumber 218-792-3
RTECSNumber AK4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only