missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylacetamide 99.0+%, TCI America™

Catalog No. p-7126226
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D046125ML 25 mL
D0461500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D046125ML Supplier TCI America Supplier No. D046125ML
Only null left

Chemical Identifiers

CAS 685-91-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00009047
InChI Key AJFDBNQQDYLMJN-UHFFFAOYSA-N
Synonym diethylacetamide, acetamide, n,n-diethyl, n-acetyldiethylamine, unii-6yo81d7st6, n,n-diaethylacetamid german, n,n-diethyl-acetamide, acetic acid, amide, n,n-diethyl, acetamide,n,n-diethyl, n,n-diaethylacetamid, n,n-diethylacetoamide
PubChem CID 12703
IUPAC Name N,N-diethylacetamide
SMILES CCN(CC)C(=O)C

Specifications

Color Colorless
Boiling Point 185°C
Quantity 25 mL
Formula Weight 115.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylacetamide
EINECSNumber (2)-3362
RTECSNumber AB7000000
TSCA Yes