missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylacetamide 99.0+%, TCI America™

Catalog No. d046125ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
D046125ML 25 mL
D0461500ML 500 mL
2 options

Catalog No. D046125ML

Supplier: TCI America D046125ML

Only null left

Chemical Identifiers

CAS 685-91-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00009047
InChI Key AJFDBNQQDYLMJN-UHFFFAOYSA-N
Synonym diethylacetamide, acetamide, n,n-diethyl, n-acetyldiethylamine, unii-6yo81d7st6, n,n-diaethylacetamid german, n,n-diethyl-acetamide, acetic acid, amide, n,n-diethyl, acetamide,n,n-diethyl, n,n-diaethylacetamid, n,n-diethylacetoamide
PubChem CID 12703
IUPAC Name N,N-diethylacetamide
SMILES CCN(CC)C(=O)C

Specifications

Color Colorless
Boiling Point 185°C
Quantity 25 mL
Formula Weight 115.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylacetamide
EINECSNumber (2)-3362
RTECSNumber AB7000000
TSCA Yes