missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylacetamide, 99%

Catalog No. AAB2076214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2076214 25 g
AAB2076222 100 g
AAB2076236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2076214 Supplier Thermo Scientific Chemicals Supplier No. B2076214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 685-91-6 | C6H13NO | 115.176 g/mol

N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylacetamide was used to investigate the spectroscopic properties ofN,N-diethyl-2-[(4-substituted)phenylsulfinyl] acetamides. It is commonly used as drug solvent.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 685-91-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00009047
InChI Key AJFDBNQQDYLMJN-UHFFFAOYSA-N
Synonym diethylacetamide, acetamide, n,n-diethyl, n-acetyldiethylamine, unii-6yo81d7st6, n,n-diaethylacetamid german, n,n-diethyl-acetamide, acetic acid, amide, n,n-diethyl, acetamide,n,n-diethyl, n,n-diaethylacetamid, n,n-diethylacetoamide
PubChem CID 12703
IUPAC Name N,N-diethylacetamide
SMILES CCN(CC)C(=O)C

Specifications

Density 0.92
Boiling Point 182°C to 186°C
Flash Point 76°C (169°F)
Refractive Index 1.44
Quantity 25 g
Beilstein 1209428
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material N,N-Diethylacetamide
Hazard Category H227-H373
Hazard Statement GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P260-P280-P314-P370+P378q-P501c
EINECSNumber 211-685-2
RTECSNumber AB7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only