missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-phenylenediamine oxalate, 96%

Catalog No. AC407410100
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407410100 10 g Glass bottle
AC407410250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407410100 Supplier Thermo Scientific Chemicals Supplier No. 407410100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62637-92-7 | 0·5 C2H2O4 | 209.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62637-92-7
Molecular Formula 0·5 C2H2O4
Molecular Weight (g/mol) 209.27
MDL Number MFCD00070242

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 145.0°C to 150.0°C
Color Brown to Gray or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 10 g
Formula Weight 209.27
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Diethyl-p-phenylenediamine oxalate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: Call a POISON CENT
EINECSNumber 263-662-1
TSCA TSCA

RUO – Research Use Only