missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-phenylenediamine, 98%

Catalog No. p-3778279
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407390050 5 g Glass bottle
AC407390250 25 g Glass bottle
AC407391000 100 g Glass bottle
AC407395000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC407390050 Supplier Thermo Scientific Chemicals Supplier No. 407390050

CAS: 93-05-0 | C10H16N2 | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-05-0
MDL Number MFCD00007861
InChI Key QNGVNLMMEQUVQK-UHFFFAOYSA-N
Synonym n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline
PubChem CID 7120
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine
SMILES CCN(CC)C1=CC=C(C=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23.0°C
Density 0.9880g/mL
Boiling Point 268.0°C
Flash Point 139°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H16N2
Linear Formula (CH3CH2)2NC6H4NH2
Refractive Index 1.5690 to 1.5730 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.988
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Molecular Weight (g/mol) 164.25
Formula Weight 164.25
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material N, N-Diethyl-p-phenylenediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 202-214-1

RUO – Research Use Only