missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-N'-methylethylenediamine 98.0+%, TCI America™

Catalog No. D129725ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D129725ML 25 mL
D12975ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D129725ML Supplier TCI America Supplier No. D129725ML
Only null left

Chemical Identifiers

CAS 104-79-0
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00009053
InChI Key MKDYQLJYEBWUIG-UHFFFAOYSA-N
Synonym n,n-diethyl-n'-methylethylenediamine, n1,n1-diethyl-n2-methylethane-1,2-diamine, 1,2-ethanediamine, n,n-diethyl-n'-methyl, 2-diethylamino-n-methylethylamine, diethyl 2-methylaminoethyl amine, 2-diethylamino ethyl methylamine, diethyl 2-methylamino ethyl amine, ethylenediamine, n,n-diethyl-n'-methyl, n,n-diethyl-n'-methylenediamine
PubChem CID 66034
IUPAC Name diethyl[2-(methylamino)ethyl]amine
SMILES CCN(CC)CCNC

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 mL
UN Number 2734
Formula Weight 130.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-N′-methylethylenediamine