missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2,2,2-trifluoroacetamide 98.0+%, TCI America™

Catalog No. p-7128637
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50635G 5 g
D506325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50635G Supplier TCI America Supplier No. D50635G

Chemical Identifiers

CAS 360-92-9
Molecular Formula C6H10F3NO
Molecular Weight (g/mol) 169.147
MDL Number MFCD00192034
InChI Key CODXZFSZJFCVBE-UHFFFAOYSA-N
Synonym n,n-diethyltrifluoroacetamide, acetamide, n,n-diethyl-2,2,2-trifluoro, acmc-1cmeq, diethyl trifluoroacetamide, n,n-diethyltrifluoro-acetamide, trifluoroacetic acid diethylamide, n,n-diethyl-2,2,2-trifluoroacetamide, n,n-diethyl-2,2,2-tris fluoranyl ethanamide
PubChem CID 225443
IUPAC Name N,N-diethyl-2,2,2-trifluoroacetamide
SMILES CCN(CC)C(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
UN Number 1993
Formula Weight 169.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-2,2,2-trifluoroacetamide