missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethyl-1,6-diaminohexane 97.0+%, TCI America™

Catalog No. p-7126669
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13055ML 5 mL
D130525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13055ML Supplier TCI America Supplier No. D13055ML

Chemical Identifiers

CAS 13093-05-5
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.32
MDL Number MFCD00026707
InChI Key LDQWVRMGQLAWMN-UHFFFAOYSA-N
PubChem CID 83132
IUPAC Name ethyl[6-(ethylamino)hexyl]amine
SMILES CCNCCCCCCNCC

Specifications

Color Yellow
Boiling Point 228°C
Quantity 5 mL
UN Number 2735
Synonym 1,6-Bis(ethylamino)hexane, N,N′C-Diethylhexamethylenediamine, N,N′C-Diethyl-1,6-hexanediamine
Formula Weight 172.32
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Diethyl-1,6-diaminohexane
EINECSNumber (2)-2374
TSCA Yes