missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. D051825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D051825G 25 g
D0518500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D051825G Supplier TCI America Supplier No. D051825G

Chemical Identifiers

CAS 93-05-0
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00007861
InChI Key QNGVNLMMEQUVQK-UHFFFAOYSA-N
Synonym n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline
PubChem CID 7120
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine
SMILES CCN(CC)C1=CC=C(C=C1)N

Specifications

Color Brown-Yellow
Boiling Point 262°C
Quantity 25 g
UN Number 2922
Formula Weight 164.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-1,4-phenylenediamine
EINECSNumber (3)-0243
RTECSNumber SS9275000
TSCA Yes
Recommended Storage Freezer