missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,4-phenylenediamine Sulfate 98.0+%, TCI America™

Catalog No. p-7126265
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D052025G 25 g
D0520100G 100 g
D0520500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D052025G Supplier TCI America Supplier No. D052025G
Only null left

Chemical Identifiers

CAS 6283-63-2
Molecular Formula C10H18N2O4S
Molecular Weight (g/mol) 262.324
MDL Number MFCD00012993
InChI Key AYLDJQABCMPYEN-UHFFFAOYSA-N
Synonym n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1
PubChem CID 80166
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid
SMILES CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O

Specifications

Melting Point 185°C
Quantity 25 g
Formula Weight 262.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethyl-1,4-phenylenediamine Sulfate
EINECSNumber (3)-0243
RTECSNumber SS9625000
TSCA No