missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,4-butanediamine, 96%

Catalog No. AAH5027806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5027806 5 g
AAH5027803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5027806 Supplier Thermo Scientific Chemicals Supplier No. H5027806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27431-62-5 | C8H20N2 | 144.262 g/mol

N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 27431-62-5
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.262
MDL Number MFCD00014836
InChI Key JILXUIANNUALRZ-UHFFFAOYSA-N
Synonym 4-aminobutyl diethylamine, 4-diethylaminobutylamine, 4-diethylamino butylamine, n1,n1-diethylbutane-1,4-diamine, n,n-diethylbutane-1,4-diamine, n,n-diethyl-1,4-butanediamine, n 1 ,n 4-diethylputrescine, 1,4-butanediamine, n,n-diethyl, diethylaminobutylamine, acmc-1cneh
PubChem CID 117976
IUPAC Name N',N'-diethylbutane-1,4-diamine
SMILES CCN(CC)CCCCN

Specifications

Density 0.83
Boiling Point 72°C to 24°C (10 mmHg)
Linear Formula (CH3CH2)2NCH2CH2CH2CH2NH2
Quantity 5 g
UN Number UN2735
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 144.26
Percent Purity 96%
Chemical Name or Material N,N-Diethyl-1,4-butanediamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 248-456-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only