missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,3-propanediamine, 99%

Catalog No. AAL13888AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13888AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL13888AP Supplier Thermo Scientific Chemicals Supplier No. L13888AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-78-9 | C7H18N2 | 130.24 g/mol

3-Diethylaminopropylamine is used as a solvent, extraction agents and organic synthesis intermediates, dyes, pigments, surface active agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Diethylaminopropylamine is used as a solvent, extraction agents and organic synthesis intermediates, dyes, pigments, surface active agent.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 104-78-9
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008218
InChI Key QOHMWDJIBGVPIF-UHFFFAOYSA-N
Synonym 3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine
PubChem CID 61011
IUPAC Name N',N'-diethylpropane-1,3-diamine
SMILES CCN(CC)CCCN

Specifications

Melting Point -60°C
Density 0.826
Boiling Point 168°C to 171°C
Flash Point 51°C (124°F)
Refractive Index 1.442
Linear Formula (CH3CH2)2N(CH2)3NH2
Quantity 500 mL
UN Number UN2684
Beilstein 741879
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material N,N-Diethyl-1,3-propanediamine
Show More Show Less
Hazard Category H226-H302+H312-H314-H317
Hazard Statement GHS H Statement
H314-H226-H302-H312-H317
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 3-DIETHYAMINO-PROPYLAMINE
EINECSNumber 203-236-4
RTECSNumber TX7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only