missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,3-diaminopropane 99.0+%, TCI America™

Catalog No. p-7126234
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D047325ML 25 mL
D0473500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D047325ML Supplier TCI America Supplier No. D047325ML
Only null left

Chemical Identifiers

CAS 104-78-9
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.24
MDL Number MFCD00008218
InChI Key QOHMWDJIBGVPIF-UHFFFAOYSA-N
Synonym 3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine
PubChem CID 61011
IUPAC Name (3-aminopropyl)diethylamine
SMILES CCN(CC)CCCN

Specifications

Color Colorless
Boiling Point 171°C
Quantity 25 mL
UN Number 2684
Formula Weight 130.24
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-1,3-diaminopropane
EINECSNumber (2)-0158
RTECSNumber TX7350000
TSCA Yes