missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine, 90%, Thermo Scientific™

Catalog No. p-3559330
Change view
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC389210250 25g Glass bottle
AC389211000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC389210250 Supplier Thermo Scientific Chemicals Supplier No. 389210250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

C-X bond formation reagent

Chemical Identifiers

CAS 309-88-6
Molecular Formula C7H11F6N
Molecular Weight (g/mol) 223.16
MDL Number MFCD00054683
InChI Key BNTFCVMJHBNJAR-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant
PubChem CID 136149
IUPAC Name N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
SMILES CCN(CC)C(C(C(F)(F)F)F)(F)F

Specifications

CAS Min % 88.0
CAS Max % 100.0
Density 1.2300g/mL
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 90% (Sum of two isomers) Approximate composition (GC)
Packaging Glass bottle
Linear Formula (CH3CH2)2NCH2F2CHFCF3
Quantity 25g
Specific Gravity 1.23
Merck Index 15, 5153
Formula Weight 223.16
Percent Purity 90%
Chemical Name or Material N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only