missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, Thermo Scientific Chemicals

Catalog No. AC155691000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155691000 100 g Glass bottle
AC155690050 5 g Glass bottle
AC155690250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC155691000 Supplier Thermo Scientific Chemicals Supplier No. 155691000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 996-50-9 | C7H19NSi | 145.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 996-50-9
Molecular Formula C7H19NSi
Molecular Weight (g/mol) 145.32
MDL Number MFCD00009040
InChI Key JOOMLFKONHCLCJ-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
PubChem CID 70454
ChEBI CHEBI:85070
IUPAC Name N-ethyl-N-trimethylsilylethanamine
SMILES CCN(CC)[Si](C)(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7600g/mL
Boiling Point 125°C to 126°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4103 to 1.4123
Linear Formula (C2H5)2NSi(CH3)3
Quantity 100 g
Fieser 03,317; 04,544; 06,634
Specific Gravity 0.76
Solubility Information Solubility in water: decomposes
Formula Weight 145.32
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 213-637-6
TSCA TSCA

RUO – Research Use Only