missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylurea, 98%

Catalog No. p-107586
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113910250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC113910250 Supplier Thermo Scientific Chemicals Supplier No. 113910250
Only null left

CAS: 2387-23-7 | C13H24N2O | 224.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2387-23-7
MDL Number MFCD00003829
InChI Key ADFXKUOMJKEIND-UHFFFAOYSA-N
Synonym n,n'-dicyclohexylurea, dicyclohexylurea, urea, n,n'-dicyclohexyl, unii-zv7823vvim, chembl1458, zv7823vvim, urea,3-dicyclohexyl, urea,n'-dicyclohexyl, n,n inverted exclamation marka-dicyclohexylurea, urea, 1,3-dicyclohexyl
PubChem CID 4277
IUPAC Name 1,3-dicyclohexylurea
SMILES C1CCC(CC1)NC(=O)NC2CCCCC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 231°C to 235°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C13H24N2O
Linear Formula C6H11NHCONHC6H11
Quantity 25 g
Beilstein 12, II, 11
Molecular Weight (g/mol) 224.35
Formula Weight 224.35
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N, N'-Dicyclohexylurea, 98%
Show More Show Less
EINECSNumber 219-213-7
TSCA TSCA

RUO – Research Use Only