missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dicyclohexylcarbodiimide 98.0+%, TCI America™
Supplier: TCI America D0436400G
Specifications
| N,N′-Dicyclohexylcarbodiimide | |
| White | |
| C13H22N2 | |
| 400 g | |
| dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl | |
| C1CCC(CC1)N=C=NC2CCCCC2 | |
| 206.333 | |
| CHEBI:53090 | |
| ≥98.0% (GC) |
| 538-75-0 | |
| 124°C | |
| MFCD00011659 | |
| 2811 | |
| QOSSAOTZNIDXMA-UHFFFAOYSA-N | |
| N,N'-dicyclohexylmethanediimine | |
| 10868 | |
| 206.33 | |
| Crystalline Lumps |
Chemical Identifiers
| 538-75-0 | |
| 206.333 | |
| QOSSAOTZNIDXMA-UHFFFAOYSA-N | |
| 10868 | |
| N,N'-dicyclohexylmethanediimine |
| C13H22N2 | |
| MFCD00011659 | |
| dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl | |
| CHEBI:53090 | |
| C1CCC(CC1)N=C=NC2CCCCC2 |
Safety and Handling
EINECSNumber : (3)-2267
RTECSNumber : FF2160000
TSCA : Yes