missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylcarbodiimide (25% in Pyridine, ca. 1.2mol/L), TCI America™

Catalog No. D0437100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0437100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0437100ML Supplier TCI America Supplier No. D0437100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 538-75-0
Molecular Formula C13H22N2
Molecular Weight (g/mol) 206.333
MDL Number MFCD00011659
InChI Key QOSSAOTZNIDXMA-UHFFFAOYSA-N
Synonym dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
PubChem CID 10868
ChEBI CHEBI:53090
IUPAC Name N,N'-dicyclohexylmethanediimine
SMILES C1CCC(CC1)N=C=NC2CCCCC2

Specifications

Color Yellow
pH 8.5 (0.2 mol H2O) (Pyridine)
Quantity 100 mL
UN Number 3286
Formula Weight 206.33
Physical Form Liquid
Chemical Name or Material N,N′-Dicyclohexylcarbodiimide (25% in Pyridine, ca. 1.2mol/L)
EINECSNumber (3)-2267
RTECSNumber FF2160000
TSCA Yes
Recommended Storage Refrigerator