missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dibutyl-3-aminophenol 95.0+%, TCI America™
$106.25 - $451.10
Chemical Identifiers
| CAS | 43141-69-1 |
|---|---|
| Molecular Formula | C14H23NO |
| Molecular Weight (g/mol) | 221.34 |
| MDL Number | MFCD00134668 |
| InChI Key | KHSTZMGCKHBFJX-UHFFFAOYSA-N |
| PubChem CID | 601234 |
| IUPAC Name | 3-(dibutylamino)phenol |
| SMILES | CCCCN(CCCC)C1=CC(O)=CC=C1 |
Chemical Identifiers
| 43141-69-1 | |
| 221.34 | |
| KHSTZMGCKHBFJX-UHFFFAOYSA-N | |
| 3-(dibutylamino)phenol |
| C14H23NO | |
| MFCD00134668 | |
| 601234 | |
| CCCCN(CCCC)C1=CC(O)=CC=C1 |
Specifications
| 43141-69-1 | |
| C14H23NO | |
| 25 g | |
| CCCCN(CCCC)C1=CC(O)=CC=C1 | |
| 221.34 | |
| 221.34 | |
| Liquid |
| 176°C | |
| MFCD00134668 | |
| KHSTZMGCKHBFJX-UHFFFAOYSA-N | |
| 3-(dibutylamino)phenol | |
| 601234 | |
| ≥95.0% (GC) | |
| N,N-Dibutyl-3-aminophenol |
Safety and Handling
EINECSNumber : (3)-3804
TSCA : No