missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibenzylhydroxylamine, 98%

Catalog No. p-7026449
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2244706 5 g
AAB2244714 25 g
AAB2244722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2244706 Supplier Thermo Scientific Chemicals Supplier No. B2244706

CAS: 621-07-8 | C14H15NO | 213.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-07-8
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00004772
InChI Key GXELTROTKVKZBQ-UHFFFAOYSA-N
Synonym dibenzylhydroxylamine, n,n-dibenzylhydroxyamine, benzenemethanamine, n-hydroxy-n-phenylmethyl, hydroxylamine, n,n-dibenzyl, bisbenzylhydroxylamine, n-benzyl-n-hydroxy-1-phenylmethanamine, acmc-1bivc, n.n-dibenzylhydroxylamine, maybridge3_004477, hydroxylamine,n-dibenzyl
PubChem CID 69297
IUPAC Name N,N-dibenzylhydroxylamine
SMILES ON(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 120°C to 125°C
Quantity 5 g
Beilstein 978234
Formula Weight 213.28
Percent Purity 98%
Chemical Name or Material N,N-Dibenzylhydroxylamine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-667-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only