missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dibenzylhydroxylamine 98.0+%, TCI America™
$155.93 - $155.93
Chemical Identifiers
| CAS | 621-07-8 |
|---|---|
| Molecular Formula | C14H15NO |
| Molecular Weight (g/mol) | 213.28 |
| MDL Number | MFCD00004772 |
| InChI Key | GXELTROTKVKZBQ-UHFFFAOYSA-N |
| Synonym | dibenzylhydroxylamine, n,n-dibenzylhydroxyamine, benzenemethanamine, n-hydroxy-n-phenylmethyl, hydroxylamine, n,n-dibenzyl, bisbenzylhydroxylamine, n-benzyl-n-hydroxy-1-phenylmethanamine, acmc-1bivc, n.n-dibenzylhydroxylamine, maybridge3_004477, hydroxylamine,n-dibenzyl |
| PubChem CID | 69297 |
| IUPAC Name | N,N-dibenzylhydroxylamine |
| SMILES | ON(CC1=CC=CC=C1)CC1=CC=CC=C1 |
Chemical Identifiers
| 621-07-8 | |
| 213.28 | |
| GXELTROTKVKZBQ-UHFFFAOYSA-N | |
| 69297 | |
| ON(CC1=CC=CC=C1)CC1=CC=CC=C1 |
| C14H15NO | |
| MFCD00004772 | |
| dibenzylhydroxylamine, n,n-dibenzylhydroxyamine, benzenemethanamine, n-hydroxy-n-phenylmethyl, hydroxylamine, n,n-dibenzyl, bisbenzylhydroxylamine, n-benzyl-n-hydroxy-1-phenylmethanamine, acmc-1bivc, n.n-dibenzylhydroxylamine, maybridge3_004477, hydroxylamine,n-dibenzyl | |
| N,N-dibenzylhydroxylamine |
Specifications
| 621-07-8 | |
| White | |
| MFCD00004772 | |
| dibenzylhydroxylamine, n,n-dibenzylhydroxyamine, benzenemethanamine, n-hydroxy-n-phenylmethyl, hydroxylamine, n,n-dibenzyl, bisbenzylhydroxylamine, n-benzyl-n-hydroxy-1-phenylmethanamine, acmc-1bivc, n.n-dibenzylhydroxylamine, maybridge3_004477, hydroxylamine,n-dibenzyl | |
| ON(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
| 213.28 | |
| 213.28 | |
| Crystalline Powder |
| 124°C | |
| C14H15NO | |
| 25 g | |
| GXELTROTKVKZBQ-UHFFFAOYSA-N | |
| N,N-dibenzylhydroxylamine | |
| 69297 | |
| ≥98.0% (HPLC,T) | |
| N,N-Dibenzylhydroxylamine |
Safety and Handling
TSCA : Yes