missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibenzylhydroxylamine 98.0+%, TCI America™

Catalog No. D200425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D200425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D200425G Supplier TCI America Supplier No. D200425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-07-8
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00004772
InChI Key GXELTROTKVKZBQ-UHFFFAOYSA-N
Synonym dibenzylhydroxylamine, n,n-dibenzylhydroxyamine, benzenemethanamine, n-hydroxy-n-phenylmethyl, hydroxylamine, n,n-dibenzyl, bisbenzylhydroxylamine, n-benzyl-n-hydroxy-1-phenylmethanamine, acmc-1bivc, n.n-dibenzylhydroxylamine, maybridge3_004477, hydroxylamine,n-dibenzyl
PubChem CID 69297
IUPAC Name N,N-dibenzylhydroxylamine
SMILES ON(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 124°C
Color White
Quantity 25 g
Formula Weight 213.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dibenzylhydroxylamine