missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzyl-3-aminopyrrolidine 97.0+%, TCI America™

Catalog No. D196425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D196425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D196425G Supplier TCI America Supplier No. D196425G
Only null left

Chemical Identifiers

CAS 108963-20-8
Molecular Formula C18H22N2
Molecular Weight (g/mol) 266.388
MDL Number MFCD00059060
InChI Key BHEXKVGLZMEJRQ-UHFFFAOYSA-N
Synonym N-Benzyl-3-(benzylamino)pyrrolidine
PubChem CID 14098325
IUPAC Name N,1-dibenzylpyrrolidin-3-amine
SMILES C1CN(CC1NCC2=CC=CC=C2)CC3=CC=CC=C3

Specifications

Color Yellow
Quantity 25 g
UN Number 2735
Formula Weight 266.39
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material N,N′-Dibenzyl-3-aminopyrrolidine