missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diaminoguanidine monohydrochloride, 98%

Catalog No. p-7026560
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
AAB2261514 25 g
AAB2261522 100 g
2 options

Catalog No. AAB2261514

Supplier: Thermo Scientific Chemicals B2261514

Only null left

CAS: 36062-19-8 | CH8ClN5 | 125.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36062-19-8
Molecular Formula CH8ClN5
Molecular Weight (g/mol) 125.56
MDL Number MFCD00012948
InChI Key HAZRIBSLCUYMQP-UHFFFAOYSA-N
Synonym 1,3-diaminoguanidine hydrochloride, unii-014y506oeg, 1,3-diaminoguanidine monohydrochloride, carbonimidic dihydrazide, hydrochloride, n,n'-diaminoguanidine monohydrochloride, n,n'-diaminoguanidine, carbonimidic dihydrazide, hydrochloride 1:?, 1,2-diaminoguanidine hydrochloride, n,n'-diaminoguandine, hydrochloride, pubchem20888
PubChem CID 9566041
IUPAC Name 1,2-diaminoguanidine;hydrochloride
SMILES C(=NN)(N)NN.Cl

Specifications

Melting Point ∼181°C (decomposition)
Quantity 25 g
Beilstein 3556702
Sensitivity Hygroscopic
Formula Weight 125.56
Percent Purity 98%
Chemical Name or Material N,N'-Diaminoguanidine monohydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 253-900-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only