missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diaminoguanidine monohydrochloride, 98%

Catalog No. AAB2261522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2261522 100 g
AAB2261514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2261522 Supplier Thermo Scientific Chemicals Supplier No. B2261522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36062-19-8 | CH8ClN5 | 125.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36062-19-8
Molecular Formula CH8ClN5
Molecular Weight (g/mol) 125.56
MDL Number MFCD00012948
InChI Key HAZRIBSLCUYMQP-UHFFFAOYSA-N
Synonym 1,3-diaminoguanidine hydrochloride, unii-014y506oeg, 1,3-diaminoguanidine monohydrochloride, carbonimidic dihydrazide, hydrochloride, n,n'-diaminoguanidine monohydrochloride, n,n'-diaminoguanidine, carbonimidic dihydrazide, hydrochloride 1:?, 1,2-diaminoguanidine hydrochloride, n,n'-diaminoguandine, hydrochloride, pubchem20888
PubChem CID 9566041
IUPAC Name 1,2-diaminoguanidine;hydrochloride
SMILES C(=NN)(N)NN.Cl

Specifications

Melting Point ∼181°C (decomposition)
Quantity 100 g
Beilstein 3556702
Sensitivity Hygroscopic
Formula Weight 125.56
Percent Purity 98%
Chemical Name or Material N,N'-Diaminoguanidine monohydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 253-900-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only