missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylhydrazine 98.0+%, TCI America™

Catalog No. D279625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D279625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D279625G Supplier TCI America Supplier No. D279625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3148-73-0
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008673
InChI Key ZLHNYIHIHQEHJQ-UHFFFAOYSA-N
Synonym n,n'-diacetylhydrazine, 1,2-diacetylhydrazine, acetic acid, 2-acetylhydrazide, diacetylahydrazine, diacetylhydrazine, sym-diacetylhydrazine, unii-ymr3zf0q4m, hydrazine, 1,2-diacetyl, 1.2-diacetylhydrazine, ymr3zf0q4m
PubChem CID 72884
ChEBI CHEBI:80616
IUPAC Name N'-acetylacetohydrazide
SMILES CC(=O)NNC(=O)C

Specifications

Melting Point 139°C
Color White
Quantity 25 g
Formula Weight 116.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diacetylhydrazine
EINECSNumber (2)-0721
TSCA Yes