missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylhydrazine, 98%

Catalog No. AAA1339330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1339330 250 g
AAA1339309 10 g
AAA1339318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1339330 Supplier Thermo Scientific Chemicals Supplier No. A1339330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3148-73-0 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3148-73-0
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008673
InChI Key ZLHNYIHIHQEHJQ-UHFFFAOYSA-N
Synonym n,n'-diacetylhydrazine, 1,2-diacetylhydrazine, acetic acid, 2-acetylhydrazide, diacetylahydrazine, diacetylhydrazine, sym-diacetylhydrazine, unii-ymr3zf0q4m, hydrazine, 1,2-diacetyl, 1.2-diacetylhydrazine, ymr3zf0q4m
PubChem CID 72884
ChEBI CHEBI:80616
IUPAC Name N'-acetylacetohydrazide
SMILES CC(=O)NNC(=O)C

Specifications

Melting Point 138°C to 142°C
Boiling Point 209°C (15 mmHg)
Quantity 250 g
Beilstein 1754018
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material N,N'-Diacetylhydrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-576-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only