missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylethylenediamine 98.0+%, TCI America™

Catalog No. D134725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D134725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D134725G Supplier TCI America Supplier No. D134725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 871-78-3
Molecular Formula C6H12N2O2
Molecular Weight (g/mol) 144.174
MDL Number MFCD00154671
InChI Key WNYIBZHOMJZDKN-UHFFFAOYSA-N
Synonym n,n'-diacetylethylenediamine, n,n'-ethylenebisacetamide, n-2-acetamidoethyl acetamide, unii-sxn3k5812f, acetamide, n,n'-1,2-ethanediylbis, nn'-diacetylethylenediamine, n-2-acetylamino ethyl acetamide, n,n'-ethylenedi diacetamide, diacetylethylenediamine, acetamide,n'-ethylenebis
PubChem CID 70097
IUPAC Name N-(2-acetamidoethyl)acetamide
SMILES CC(=O)NCCNC(=O)C

Specifications

Melting Point 175°C
Color White
Quantity 25 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diacetylethylenediamine