missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylchitobiose 98.0+%, TCI America™

Catalog No. D421520MG
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D421520MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D421520MG Supplier TCI America Supplier No. D421520MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35061-50-8
Molecular Formula C16H28N2O11
Molecular Weight (g/mol) 424.403
MDL Number MFCD00077715
InChI Key CDOJPCSDOXYJJF-FWQAMMJLSA-N
Synonym 2-Acetamido-2-deoxy-4-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-D-glucopyranose, GlcNAc beta(1-4)GlcNAc
PubChem CID 133612055
IUPAC Name N-[5-[(4S)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2,4-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1O)CO)OC2C(C(C(C(O2)CO)O)O)NC(=O)C)O

Specifications

Melting Point 247°C
Color White
Quantity 20 mg
Formula Weight 424.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Diacetylchitobiose
TSCA No
Recommended Storage Freezer