missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-tert-butylethylenediamine 98.0+%, TCI America™

Catalog No. p-7127416
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D263825ML 25 mL
D2638500ML 500 mL
2 options

Catalog No. D263825ML

Supplier: TCI America D263825ML

Only null left

Chemical Identifiers

CAS 4062-60-6
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.316
MDL Number MFCD00014996
InChI Key KGHYGBGIWLNFAV-UHFFFAOYSA-N
Synonym n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine
PubChem CID 77680
IUPAC Name N,N'-ditert-butylethane-1,2-diamine
SMILES CC(C)(C)NCCNC(C)(C)C

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 mL
UN Number 2735
Formula Weight 172.32
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Di-tert-butylethylenediamine
RTECSNumber KH8587137
TSCA Yes