missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-tert-butylethylenediamine 98.0+%, TCI America™

Catalog No. D2638500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2638500ML 500 mL
D263825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2638500ML Supplier TCI America Supplier No. D2638500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4062-60-6
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.316
MDL Number MFCD00014996
InChI Key KGHYGBGIWLNFAV-UHFFFAOYSA-N
Synonym n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine
PubChem CID 77680
IUPAC Name N,N'-ditert-butylethane-1,2-diamine
SMILES CC(C)(C)NCCNC(C)(C)C

Specifications

Color Yellow
Boiling Point 198°C
Quantity 500 mL
UN Number 2735
Formula Weight 172.32
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N′-Di-tert-butylethylenediamine
RTECSNumber KH8587137
TSCA Yes