missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-tert-butylethylenediamine, 98%

CAS: 4062-60-6 | C10H24N2 | 172.316 g/mol

Supplier:  Thermo Scientific Chemicals L0497214

 View more versions of this product

Catalog No. AAL0497214

Only null left
Explore available promotions

Description

Description

N,N'-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

Solubility
Soluble (33 g/L) (25°C),

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
Specifications

Specifications

N,N'-Di-tert-butylethylenediamine
0.801
62°C (143°F)
1.43
MFCD00014996
UN2735
n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine
KGHYGBGIWLNFAV-UHFFFAOYSA-N
N,N'-ditert-butylethane-1,2-diamine
77680
98%
4062-60-6
132°C (100 mmHg)
C10H24N2
(CH3)3CNHCH2CH2NHC(CH3)3
25 g
1738432
Soluble (33g/L) (25°C),
CC(C)(C)NCCNC(C)(C)C
172.316
172.32

Chemical Identifiers

4062-60-6
172.316
KGHYGBGIWLNFAV-UHFFFAOYSA-N
77680
CC(C)(C)NCCNC(C)(C)C
C10H24N2
MFCD00014996
n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine
N,N'-ditert-butylethane-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 223-769-6

RTECSNumber : KH8587137

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only