missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Di-sec-butyl-1,4-phenylenediamine 98.0+%, TCI America™
$106.25 - $570.40
Chemical Identifiers
| CAS | 101-96-2 |
|---|---|
| Molecular Formula | C14H24N2 |
| Molecular Weight (g/mol) | 220.36 |
| MDL Number | MFCD00043658 |
| InChI Key | FSWDLYNGJBGFJH-UHFFFAOYSA-N |
| PubChem CID | 7589 |
| IUPAC Name | 1-N,4-N-di(butan-2-yl)benzene-1,4-diamine |
| SMILES | CCC(C)NC1=CC=C(C=C1)NC(C)CC |
Chemical Identifiers
| 101-96-2 | |
| 220.36 | |
| FSWDLYNGJBGFJH-UHFFFAOYSA-N | |
| 1-N,4-N-di(butan-2-yl)benzene-1,4-diamine |
| C14H24N2 | |
| MFCD00043658 | |
| 7589 | |
| CCC(C)NC1=CC=C(C=C1)NC(C)CC |
Specifications
| 101-96-2 | |
| C14H24N2 | |
| 25 mL | |
| FSWDLYNGJBGFJH-UHFFFAOYSA-N | |
| 1-N,4-N-di(butan-2-yl)benzene-1,4-diamine | |
| 7589 | |
| ≥98.0% (GC) | |
| N,N′-Di-sec-butyl-1,4-phenylenediamine |
| 159°C | |
| MFCD00043658 | |
| 2922 | |
| CCC(C)NC1=CC=C(C=C1)NC(C)CC | |
| 220.36 | |
| 220.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0247
RTECSNumber : SS9040000
TSCA : Yes