missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Di-sec-butyl-1,4-phenylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 159°C |
| Quantity | 500 mL |
| UN Number | 2922 |
| Formula Weight | 220.36 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | N,N′-Di-sec-butyl-1,4-phenylenediamine |
Safety and Handling
| EINECSNumber | (3)-0247 |
| RTECSNumber | SS9040000 |
| TSCA | Yes |