missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-n-octyl-3,4,9,10-perylenetetracarboxylic Diimide 98.0+%, TCI America™

Catalog No. D41751G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41751G Supplier TCI America Supplier No. D41751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78151-58-3
Molecular Formula C40H42N2O4
Molecular Weight (g/mol) 614.786
MDL Number MFCD08276854
InChI Key YFGMQDNQVFJKTR-UHFFFAOYSA-N
Synonym PTCDI-C8
PubChem CID 10722480
SMILES CCCCCCCCN1C(=O)C2=C3C(=CC=C4C3=C(C=C2)C5=C6C4=CC=C7C6=C(C=C5)C(=O)N(C7=O)CCCCCCCC)C1=O

Specifications

Quantity 1 g
Formula Weight 614.79
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di-n-octyl-3,4,9,10-perylenetetracarboxylic Diimide