missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-n-butylurea, 98%

Catalog No. AAL0616506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0616506 5 g
AAL0616514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0616506 Supplier Thermo Scientific Chemicals Supplier No. L0616506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1792-17-2 | C9H20N2O | 172.27 g/mol

N,N'-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1792-17-2
Molecular Formula C9H20N2O
Molecular Weight (g/mol) 172.27
MDL Number MFCD00015229
InChI Key AQSQFWLMFCKKMG-UHFFFAOYSA-N
Synonym n,n'-dibutylurea, n,n'-di-n-butylurea, urea, n,n'-dibutyl, 1,3-dibutyl urea, unii-j2hug1i10w, j2hug1i10w, n-butyl butylamino carboxamide, nn'-dibutylurea, urea,n'-dibutyl, 1,3-dibutyl-urea
PubChem CID 74523
IUPAC Name 1,3-dibutylurea
SMILES CCCCNC(=O)NCCCC

Specifications

Melting Point 69°C to 75°C
Boiling Point 115°C (0.03 mmHg)
Quantity 5 g
Beilstein 1704165
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material N,N'-Di-n-butylurea
EINECSNumber 217-258-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only