missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%

CAS: 31687-58-8 | C28H48N4O6 | 536.714 g/mol

$115.99 - $342.47

Chemical Identifiers

CAS 31687-58-8
Molecular Formula C28H48N4O6
Molecular Weight (g/mol) 536.714
InChI Key WBGMQHNUPJENDC-MERQFXBCSA-N
Synonym boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
PubChem CID 16211319
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6201803
View Documents
Thermo Scientific Chemicals
H6201803
1 g N/A
Only null left
 
AAH6201806
View Documents
Thermo Scientific Chemicals
H6201806
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

31687-58-8
536.714
boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
C28H48N4O6
WBGMQHNUPJENDC-MERQFXBCSA-N
16211319
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

160°C to 165°C (decomposition)
White
boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2
1 g
16211319
95%
N,N'-Di-Boc-L-histidine dicyclohexylammonium salt
31687-58-8
C28H48N4O6
WBGMQHNUPJENDC-MERQFXBCSA-N
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid
536.714
536.7
Solid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 250-764-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only